APO455834030 Apollo Scientific Ltd

2,6-Difluoroanisole

  • MDL number MFCD00142846
  • Cas Number : 437-82-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.221
Flash Point 42
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 70-72/56mm
Product Link https://store.apolloscientific.co.uk/product/26-difluoroanisole

Safety Information

signalword Warning
pcodes P210;P261;P280
MSDS https://store.apolloscientific.co.uk/product/26-difluoroanisole
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart