APO455834034 Apollo Scientific Ltd

1-Bromo-1,1,2,2-tetrafluoropentane

  • MDL number MFCD00153716
  • Cas Number : 151831-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=617

Safety Information

signalword Warning
pcodes P280;P210
MSDS http://www.apolloscientific.co.uk/display_item.php?id=617
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart