APO455834038 Apollo Scientific Ltd

4-Chloro-2-(trifluoromethyl)benzaldehyde

  • MDL number MFCD03092987
  • Cas Number : 320-43-4
  • Molecular Weight : 208.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Flash Point 86
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-chloro-2-trifluoromethylbenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-2-trifluoromethylbenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart