APO455834050 Apollo Scientific Ltd

3'-Fluoro-4'-hydroxyacetophenone

  • MDL number MFCD00215840
  • Cas Number : 403-14-5
  • Molecular Weight : 154.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127-129
Synonym 1-(3-Fluoro-4-hydroxyphenyl)ethan-1-one, 4-Acetyl-2-fluorophenol
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-4-hydroxyacetophenone

Safety Information

signalword Warning
pcodes P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-4-hydroxyacetophenone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart