APO455834064 Apollo Scientific Ltd

5-Cyano-2-fluorobenzeneboronic acid

  • MDL number MFCD03095147
  • Cas Number : 468718-30-1
  • Molecular Weight : 164.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 300
Synonym 3-Borono-4-fluorobenzonitrile
Purity 98
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-cyano-2-fluorobenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-cyano-2-fluorobenzeneboronic-acid
pictograms GHS07
hstatements H302+H312+H332;H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart