APO455834066 Apollo Scientific Ltd

3-Chloro-2-fluoronitrobenzene

  • MDL number MFCD00069417
  • Cas Number : 2106-49-2
  • Molecular Weight : 175.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Chloro-2-fluoro-3-nitrobenzene
Purity 98
Flash Point >110
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 106/5mm
Product Link https://store.apolloscientific.co.uk/product/3-chloro-2-fluoronitrobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-2-fluoronitrobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart