APO455834069 Apollo Scientific Ltd

2,2-Difluoro-1,3-dimethylimidazolidine

  • MDL number MFCD03788478
  • Cas Number : 220405-40-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -8.7
Density 1.096
Flash Point 40
Storage temp. Ambient Temperature
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 47/37mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=6277

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=6277
pictograms GHS02;GHS05;GHS06
hstatements H226;H301+H311+H331;H314
Availability details for
Quantity
  • Availability
  • Add to Cart