APO455834073 Apollo Scientific Ltd

4-Fluorobenzene-1,2-diol

  • MDL number MFCD01317539
  • Cas Number : 367-32-8
  • Molecular Weight : 128.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90-91
Synonym 4-Fluorocatechol, 1,2-Dihydroxy-4-fluorobenzene
Purity 97
Flash Point >110
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-fluorobenzene-12-diol

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluorobenzene-12-diol
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart