APO455834078 Apollo Scientific Ltd

Ethyl 4,4-difluorocyclohexane-1-carboxylate

  • MDL number MFCD03788494
  • Cas Number : 178312-47-5
  • Molecular Weight : 192.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4,4-Difluoro-1-(ethoxycarbonyl)cyclohexane
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 84-86/8mm
Product Link https://store.apolloscientific.co.uk/product/ethyl-44-difluorocyclohexane-1-carboxylate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-44-difluorocyclohexane-1-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart