APO455834085 Apollo Scientific Ltd

2,4-Dibromo-6-fluorobenzaldehyde

  • MDL number MFCD03094089
  • Cas Number : 205683-34-7
  • Molecular Weight : 281.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 57-60
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/24-dibromo-6-fluorobenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/24-dibromo-6-fluorobenzaldehyde
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart