APO455834090 Apollo Scientific Ltd

1-Bromo-1,1,2,2-tetrafluorooctane

  • MDL number MFCD00153714
  • Cas Number : 231630-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 1.7
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 67-69/20mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2512

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=2512
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart