APO455834097 Apollo Scientific Ltd

3-Iodophenylsulphur pentafluoride

  • MDL number MFCD03788513
  • Cas Number : 286947-67-9
  • Molecular Weight : 330.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-iodo-3-(pentafluoro-lambda~6~-sulfanyl)benzene
density 2.06
Inchi Code 1S/C6H4F5IS/c7-13(8,9,10,11)6-3-1-2-5(12)4-6/h1-4H
Inchi Key OPPXUPGOUVHFHM-UHFFFAOYSA-N

Description

Synonym 1-Iodo-3-(pentafluorothio)benzene, 1-Iodo-3-(pentafluorosulphanyl)benzene
Density 2.06
Flash Point 57/0.75mm
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-iodophenylsulphur-pentafluoride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-iodophenylsulphur-pentafluoride
pictograms GHS07
hstatements H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart