APO455834100 Apollo Scientific Ltd

3-(Trifluoromethyl)dihydrofuran-2(3H)-one

  • MDL number MFCD00798462
  • Cas Number : 174744-18-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Oxo-3-(trifluoromethyl)tetrahydrofuran, 3-(Trifluoromethyl)oxolan-2-one, alpha-(Trifluoromethyl)-gamma-butyrolactone
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=11498

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=11498
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart