APO455834101 Apollo Scientific Ltd

4-Hydroxyphenylsulphur pentafluoride

  • MDL number MFCD03788516
  • Cas Number : 774-94-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104-107
Synonym 4-(Pentafluorothio)phenol, 4-(Pentafluorosulphanyl)phenol
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=6343

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=6343
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart