APO455834103 Apollo Scientific Ltd

3-(Pentafluorothio)benzaldehyde

  • MDL number MFCD03788517
  • Cas Number : 401892-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Formylphenylsulphur pentafluoride, 3-(Pentafluorosulphanyl)benzaldehyde
Purity 97
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=6344

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=6344
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart