APO455834104 Apollo Scientific Ltd

3,5-Dichlorobenzotrifluoride

  • MDL number MFCD00203929
  • Cas Number : 54773-20-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -1
Synonym 1,3-Dichloro-5-(trifluoromethyl)benzene, 3,5-Dichloro-alpha,alpha,alpha-trifluorotoluene
Density 1.47
Flash Point 65
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 173-174
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2963

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=2963
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart