APO455834105 Apollo Scientific Ltd

4-(Pentafluorothio)benzaldehyde

  • MDL number MFCD03788518
  • Cas Number : 401892-84-0
  • Molecular Weight : 232.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Formylphenylsulphur pentafluoride, 4-(Pentafluorosulphanyl)benzaldehyde
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-pentafluorothiobenzaldehyde

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-pentafluorothiobenzaldehyde
pictograms GHS06
hstatements H301+H311+H331
Availability details for
Quantity
  • Availability
  • Add to Cart