APO455834106 Apollo Scientific Ltd

3-(Pentafluorothio)benzoic acid

  • MDL number MFCD03788519
  • Cas Number : 833-96-5
  • Molecular Weight : 248.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-162
Synonym 3-Carboxyphenylsulphur pentafluoride, 3-(Pentafluorosulphanyl)benzoic acid
Purity 95
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-pentafluorothiobenzoic-acid

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-pentafluorothiobenzoic-acid
pictograms GHS06
hstatements H301+H311+H331
Availability details for
Quantity
  • Availability
  • Add to Cart