APO455834109 Apollo Scientific Ltd

3-(Pentafluorothio)benzonitrile

  • MDL number MFCD03788520
  • Cas Number : 401892-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Cyanophenylsulphur pentafluoride, 3-(Pentafluorosulphanyl)benzonitrile
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=6348

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=6348
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart