APO455834110 Apollo Scientific Ltd

4-(Pentafluorothio)benzonitrile

  • MDL number MFCD03788521
  • Cas Number : 401892-85-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102-105
Synonym 4-Cyanophenylsulphur pentafluoride, 4-(Pentafluorosulphanyl)benzonitrile
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=6349

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=6349
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart