APO455834113 Apollo Scientific Ltd

4-(Pentafluorosulphanyl)benzoyl chloride

  • MDL number MFCD03788523
  • Cas Number : 197384-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(Pentafluorothio)benzoyl chloride, 4-(Chlorocarbonyl)phenylsulphur pentafluoride
Purity 97
Storage temp. Ambient Temperature
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=6351

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=6351
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart