APO455834114 Apollo Scientific Ltd

2,4-Dinitro-4'-fluorodiphenyl ether

  • MDL number MFCD01046749
  • Cas Number : 1033-02-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,4-Dinitro-1-(4-fluorophenoxy)benzene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=6352

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=6352
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart