APO455834117 Apollo Scientific Ltd

Acetone 2-nitro-4-(trifluoromethyl)phenylhydrazone

  • MDL number MFCD00763903
  • Cas Number : 99422-00-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92-93
Synonym N-Isopropylidene-N'-[2-nitro-4-(trifluoromethyl)phenyl]hydrazine, N-[2-Nitro-4-(trifluoromethyl)phenyl]-N'-(prop-2-ylidene)hydrazine
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=6355

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=6355
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart