APO455834121 Apollo Scientific Ltd

5-Fluoro-2-nitroanisole

  • MDL number MFCD00077541
  • Cas Number : 448-19-1
  • Molecular Weight : 171.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49
Synonym 4-Fluoro-2-methoxy-1-nitrobenzene, 5-Fluoro-2-nitrophenyl methyl ether
Purity 99
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 272
Product Link https://store.apolloscientific.co.uk/product/5-fluoro-2-nitroanisole

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-fluoro-2-nitroanisole
pictograms GHS07;GHS08
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart