APO455834122 Apollo Scientific Ltd

3-Fluoro-2-methoxybenzoic acid

  • MDL number MFCD00973972
  • Cas Number : 106428-05-1
  • Molecular Weight : 170.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112-114
Flash Point >110
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-2-methoxybenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-2-methoxybenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart