APO455834132 Apollo Scientific Ltd

3,5-Difluorobenzyl chloride

  • MDL number MFCD01090990
  • Cas Number : 220141-71-9
  • Molecular Weight : 162.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(chloromethyl)-3,5-difluorobenzene
Inchi Code 1S/C7H5ClF2/c8-4-5-1-6(9)3-7(10)2-5/h1-3H,4H2
Inchi Key VNGSMSFVLAAOGK-UHFFFAOYSA-N

Description

Synonym 1-(Chloromethyl)-3,5-difluorobenzene, alpha-Chloro-3,5-difluorotoluene
Purity 98%
Flash Point 82/22mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/35-difluorobenzyl-chloride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/35-difluorobenzyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart