APO455834134 Apollo Scientific Ltd

2-Chloro-4-fluoro-5-nitroanisole

  • MDL number MFCD03425810
  • Cas Number : 84478-76-2
  • Molecular Weight : 205.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72-75
Synonym 1-Chloro-5-fluoro-2-methoxy-4-nitrobenzene, 2-Chloro-4-fluoro-5-nitrophenyl methyl ether
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-4-fluoro-5-nitroanisole

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-4-fluoro-5-nitroanisole
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart