APO455834138 Apollo Scientific Ltd

Methyl 2-fluoro-6-methoxybenzoate

  • MDL number MFCD03788537
  • Cas Number : 178747-79-0
  • Molecular Weight : 184.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp >110
Iupac Name methyl 2-fluoro-6-methoxybenzoate
Inchi Code 1S/C9H9FO3/c1-12-7-5-3-4-6(10)8(7)9(11)13-2/h3-5H,1-2H3
Inchi Key WOKPPDUGASITJX-UHFFFAOYSA-N

Description

Melting Point >110
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-2-fluoro-6-methoxybenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2-fluoro-6-methoxybenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart