APO455834139 Apollo Scientific Ltd

2-Fluoro-6-methoxyaniline

  • MDL number MFCD03788538
  • Cas Number : 446-61-7
  • Molecular Weight : 141.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Amino-3-fluoroanisole, 6-Fluoro-o-anisidine
Flash Point 68
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 208/756mm
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-6-methoxyaniline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-6-methoxyaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart