APO455834141 Apollo Scientific Ltd

Hexakis(2,2,3,3-tetrafluoropropoxy)phosphazene

  • MDL number MFCD00798140
  • Cas Number : 58943-98-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,2,4,4,6,6-Hexakis(2,2,3,3-tetrafluoropropoxy)-1,3,5-triazatriphosphine, Hexakis(1H,1H,3H-perfluoropropoxy)phosphazene, Hexakis(1H,1H,3H-tetrafluoropropoxy)phosphazene
Storage temp. Ambient Temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 174-176/2mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=3677

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=3677
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart