APO455834142 Apollo Scientific Ltd

2-Bromo-3-fluoroanisole

  • MDL number MFCD03788540
  • Cas Number : 446-59-3
  • Molecular Weight : 205.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromo-1-fluoro-3-methoxybenzene, 2-Bromo-3-fluorophenyl methyl ether
Purity 99
Flash Point 81
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 220
Product Link https://store.apolloscientific.co.uk/product/2-bromo-3-fluoroanisole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-3-fluoroanisole
pictograms GHS07
hstatements H315;H319;H335;H412
Availability details for
Quantity
  • Availability
  • Add to Cart