APO455834151 Apollo Scientific Ltd

3-Fluoro-2-methylbenzyl bromide

  • MDL number MFCD03788543
  • Cas Number : 500912-14-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(Bromomethyl)-3-fluoro-2-methylbenzene
Purity 98
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=6370

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=6370
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart