APO455834153 Apollo Scientific Ltd

3-Fluoro-4-(trifluoromethyl)benzonitrile

  • MDL number MFCD00236280
  • Cas Number : 231953-38-1
  • Molecular Weight : 189.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 45-48
Synonym 4-Cyano-2-fluorobenzotrifluoride, alpha,alpha,alpha,3-Tetrafluoro-p-tolunitrile
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 194
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-4-trifluoromethylbenzonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-4-trifluoromethylbenzonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart