APO455834154 Apollo Scientific Ltd

2-Fluoro-6-methoxybenzyl alcohol

  • MDL number MFCD00671766
  • Cas Number : 253668-46-1
  • Molecular Weight : 156.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 35-39
Synonym (2-Fluoro-6-methoxyphenyl)methanol, 3-Fluoro-2-(hydroxymethyl)anisole
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-6-methoxybenzyl-alcohol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-6-methoxybenzyl-alcohol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart