APO455834156 Apollo Scientific Ltd

1-Chloro-1,2-dibromotrifluoroethane

  • MDL number MFCD00039316
  • Cas Number : 354-51-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -72.9
Synonym 1-Chloro-1,2-dibromo-1,2,2-trifluoroethane
Purity 98
Density 2.248
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 93-94
Product Link http://www.apolloscientific.co.uk/display_item.php?id=734

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=734
pictograms GHS07
hstatements H315;H319;H335;EUH059
Availability details for
Quantity
  • Availability
  • Add to Cart