APO455834157 Apollo Scientific Ltd

2-Fluoro-6-methoxybenzyl bromide

  • MDL number MFCD08688579
  • Cas Number : 500912-17-4
  • Molecular Weight : 219.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 18-21
Iupac Name 2-(bromomethyl)-3-fluorophenyl methyl ether
Inchi Code 1S/C8H8BrFO/c1-11-8-4-2-3-7(10)6(8)5-9/h2-4H,5H2,1H3
Inchi Key PVKFBCBTHHTDEX-UHFFFAOYSA-N

Description

Synonym 2-(Bromomethyl)-3-fluoroanisole, 2-(Bromomethyl)-1-fluoro-3-methoxybenzene, 2-(Bromomethyl)-3-fluorophenyl methyl ether
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 18-21
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-6-methoxybenzyl-bromide

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-6-methoxybenzyl-bromide
pictograms GHS05;GHS07
hstatements H314;H335
Availability details for
Quantity
  • Availability
  • Add to Cart