APO455834158 Apollo Scientific Ltd

2'-Chloro-6'-fluoroacetophenone

  • MDL number MFCD01631390
  • Cas Number : 87327-69-3
  • Molecular Weight : 172.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(2-Chloro-6-fluorophenyl)ethan-1-one
Purity 97
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-6-fluoroacetophenone

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-6-fluoroacetophenone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart