APO455834163 Apollo Scientific Ltd

2H-Perfluoro(5,8-dimethyl-3,6,9-trioxadodecane)

  • MDL number MFCD00054715
  • Cas Number : 3330-16-3
  • Molecular Weight : 618.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp -106.7
Iupac Name 1-{1-[difluoro(1,2,2,2-tetrafluoroethoxy)methyl]-1,2,2,2-tetrafluoroethoxy}-1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propane
density 1.723
Inchi Code 1S/C11HF23O3/c12-1(2(13,14)15)35-10(31,32)4(18,7(23,24)25)37-11(33,34)5(19,8(26,27)28)36-9(29,30)3(16,17)6(20,21)22/h1H
Inchi Key QRLOKTBZZPZAMX-UHFFFAOYSA-N

Description

Purity 97%
Density 1.723
Flash Point 151-154
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point -106.7
Product Link https://store.apolloscientific.co.uk/product/2h-perfluoro58-dimethyl-369-trioxadodecane

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2h-perfluoro58-dimethyl-369-trioxadodecane
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart