APO455834165 Apollo Scientific Ltd

3,5-Difluoro-2-nitrobenzoic acid

  • MDL number MFCD04035649
  • Cas Number : 331765-71-0
  • Molecular Weight : 203.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Carboxy-4,6-difluoronitrobenzene
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/35-difluoro-2-nitrobenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/35-difluoro-2-nitrobenzoic-acid
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart