APO455834175 Apollo Scientific Ltd

2-[(2-Fluorophenyl)amino]benzoic acid

  • MDL number MFCD00662257
  • Cas Number : 54-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 186
Synonym N-(2-Fluorophenyl)anthranilic acid, 2-(2-Fluoroanilino)benzoic acid
Purity 98
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9483

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=9483
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart