APO455834182 Apollo Scientific Ltd

3-Bromo-4-fluorophenol

  • MDL number MFCD03425884
  • Cas Number : 27407-11-0
  • Molecular Weight : 191

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Flash Point 99
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 78/0.5mm
Product Link https://store.apolloscientific.co.uk/product/3-bromo-4-fluorophenol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-4-fluorophenol
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart