APO455834188 Apollo Scientific Ltd

1-[2-Fluoro-4-(methylsulphonyl)phenyl]homopiperazine

  • MDL number MFCD06800757
  • Cas Number : 849924-88-5
  • Molecular Weight : 272.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(1,4-Diazepan-1-yl)-3-fluorophenyl methyl sulphone, 1-[2-Fluoro-4-(methylsulphonyl)phenyl]-1,4-diazepane
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-2-fluoro-4-methylsulphonylphenylhomopiperazine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-2-fluoro-4-methylsulphonylphenylhomopiperazine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart