APO455834192 Apollo Scientific Ltd

5-Hydroxy-2-(trifluoromethyl)pyridine

  • MDL number MFCD06801325
  • Cas Number : 216766-12-0
  • Molecular Weight : 163.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173-176(dec.)
Synonym 2-(Trifluoromethyl)pyridin-5-ol, 6-(Trifluoromethyl)pyridin-3-ol
Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-hydroxy-2-trifluoromethylpyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-hydroxy-2-trifluoromethylpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart