APO455834193 Apollo Scientific Ltd

(E)-3-[6-(Trifluoromethyl)pyridin-3-yl]acrylic acid

  • MDL number MFCD06205688
  • Cas Number : 944805-93-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 232-236
Synonym (2E)-3-[6-(Trifluoromethyl)pyridin-3-yl]prop-2-enoic acid, trans-3-[6-(Trifluoromethyl)pyridin-3-yl]acrylic acid
Purity 95
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9508

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=9508
pictograms GHS07
hstatements H302;H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart