APO455834200 Apollo Scientific Ltd

4-Amino-3-fluorobenzonitrile

  • MDL number MFCD00055559
  • Cas Number : 63069-50-1
  • Molecular Weight : 136.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87-90
Synonym 4-Cyano-2-fluoroaniline
Purity 98
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-amino-3-fluorobenzonitrile

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-3-fluorobenzonitrile
pictograms GHS06
hstatements H301+H311+H331;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart