APO455834201 Apollo Scientific Ltd

2,4-Difluoro-5-nitrobenzonitrile

  • MDL number MFCD07772030
  • Cas Number : 67152-20-9
  • Molecular Weight : 184.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Cyano-2,4-difluoronitrobenzene
Purity 98
Flash Point >110
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/24-difluoro-5-nitrobenzonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/24-difluoro-5-nitrobenzonitrile
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart