APO455834204 Apollo Scientific Ltd

5-Phenyl-3-(trifluoromethyl)-1H-pyrazole

  • MDL number MFCD00156064
  • Cas Number : 4027-54-7
  • Molecular Weight : 212.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121
Synonym 3-Phenyl-5-(trifluoromethyl)-1H-pyrazole
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-phenyl-3-trifluoromethyl-1h-pyrazole

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-phenyl-3-trifluoromethyl-1h-pyrazole
pictograms GHS06
hstatements H301;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart