APO455834210 Apollo Scientific Ltd

1,2,4-Trichloro-1,1,2-trifluorobutane

  • MDL number MFCD00077597
  • Cas Number : 261760-97-8
  • Molecular Weight : 215.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 134-136
Product Link https://store.apolloscientific.co.uk/product/124-trichloro-112-trifluorobutane

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/124-trichloro-112-trifluorobutane
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart