APO455834214 Apollo Scientific Ltd

(4-Methoxybenzoyl)-1,1,1-trifluoroacetone

  • MDL number MFCD03238723
  • Cas Number : 15191-68-1
  • Molecular Weight : 246.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 57-58
Iupac Name (2Z)-4,4,4-trifluoro-3-hydroxy-1-(4-methoxyphenyl)-2-buten-1-one
Inchi Code 1S/C11H9F3O3/c1-17-8-4-2-7(3-5-8)9(15)6-10(16)11(12,13)14/h2-5H,6H2,1H3
Inchi Key NNEIYSHJFCLFES-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 57-58
Product Link https://store.apolloscientific.co.uk/product/4-methoxybenzoyl-111-trifluoroacetone

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-methoxybenzoyl-111-trifluoroacetone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart