APO455834217 Apollo Scientific Ltd

Diethyl 2-(4-fluorophenyl)malonate

  • MDL number MFCD04039208
  • Cas Number : 2965-90-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Diethyl 2-(4-fluorophenyl)propane-1,3-dioate
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 161-162/15mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=8634

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=8634
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart